Methyl-2,4-dimethoxy-6-methylbenzoate 98%

L09890-MD L09890-03 L09890-03 L09890-MD
AAAL09890-03EA 163.02 USD
AAAL09890-03 CAAAAL09890-03 AAAL09890-MD CAAAAL09890-MD
Methyl-2,4-dimethoxy-6-methylbenzoate 98%
Methyl-2,4-dimethoxy-6-methylbenzoate
Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00082782
CAS Number: 6110-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR