1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione 97%
|
Danger
|
L09724-06
L09724-14
L09724-14
L09724-03
L09724-03
L09724-06
AAAL09724-14EA
519.48
USD
AAAL09724-14
AAAL09724-03
CAAAAL09724-14
CAAAAL09724-03
AAAL09724-06
CAAAAL09724-06
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione 97%
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
Formula:
C₇H₂F₁₀O₂ MW: 308.08 Boiling Pt: 98…100 °C Flash Pt: None Storage Temperature: Ambient |
UN:
3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...