D-Phenylalaninol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: D-PenylalaninolD(+)-2-Amino-3-phenyl-1-propanol

L09697-03 L09697-03
AAAL09697-03EA 46.89 USD
AAAL09697-03 CAAAAL09697-03
D-Phenylalaninol 98%
D-Phenylalaninol
Formula: C₉H₁₃NO
MW: 151.2 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00064298
CAS Number: 5267-64-1
EINECS: 226-086-1
UN: 3259
ADR: "8",III

Order Now

Specification Test Results

Optical rotation +23° (c=5 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR