4-(4-Fluorophenyl)-3-thiosemicarbazide 98%

Supplier: Thermo Scientific Chemicals

Danger

L09482-06 L09482-06
AAAL09482-06EA 132 USD
CAAAAL09482-06 AAAL09482-06
4-(4-Fluorophenyl)-3-thiosemicarbazide 98%
4-(4-Fluorophenyl)-3-thiosemicarbazide
Formula: C₇H₈FN₃S
MW: 185.22
Melting Pt: ca 179 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041294
CAS Number: 330-94-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR