(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-1-Benzyl-3-aminopyrrolidine
, (S)-(+)-1-Benzyl-3-aminopyrrolidine
L09449-03
L09449-06
L09449-03
L09449-06
AAAL09449-03EA
116.99
USD
AAAL09449-03
CAAAAL09449-03
AAAL09449-06
CAAAAL09449-06
(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Boiling Pt: 100…105 °C/3 mm Density: 1.020 Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082766 CAS Number: 114715-38-7 UN: 2735 ADR: "8",III |
Specification Test Results
Optical rotation | +5° (c=10 in ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...