4-(2-Methylphenyl)-3-thiosemicarbazide 98%
|
Danger
|
L09382-06
L09382-14
L09382-14
L09382-06
AAAL09382-14EA
147.63
USD
AAAL09382-14
CAAAAL09382-14
AAAL09382-06
CAAAAL09382-06
4-(2-Methylphenyl)-3-thiosemicarbazide 98%
4-(2-Methylphenyl)-3-thiosemicarbazide
Formula:
C₈H₁₁N₃S Melting Pt: 145…148 °C Storage Temperature: Ambient |
UN:
2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...