1-(2-(Trifluoromethyl)phenyl)thiourea 98%

Danger

Synonyms: N-[2-(Trifluoromethyl)phenyl]thiourea

L09377-06 L09377-03 L09377-03 L09377-06
AAAL09377-06EA 123.21 USD
AAAL09377-06 CAAAAL09377-06 AAAL09377-03 CAAAAL09377-03
1-(2-(Trifluoromethyl)phenyl)thiourea 98%
1-(2-(Trifluoromethyl)phenyl)thiourea
Formula: C₈H₇F₃N₂S
MW: 220.22
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00041161
CAS Number: 1736-71-6
EINECS: 217-084-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR