Propargyl propionate 98%
|
Warning
|
L09371-18
L09371-09
L09371-18
L09371-09
AAAL09371-18EA
209.88
USD
AAAL09371-18
CAAAAL09371-18
AAAL09371-09
CAAAAL09371-09
Propargyl propionate 98%
Propargyl propionate
Formula:
C₆H₈O₂ Boiling Pt: 137…139 °C Density: 0.973 Flash Pt: 40 °C(104 °F) Storage Temperature: Ambient |
EINECS:
217-695-3 UN: 3272 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...