Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate 97%
|
Warning
|
L09368-03
L09368-03
L09368-06
L09368-06
AAAL09368-03EA
49.45
USD
CAAAAL09368-03
AAAL09368-03
CAAAAL09368-06
AAAL09368-06
Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate 97%
Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate
Formula:
C₁₀H₉NO₂S Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053052 CAS Number: 74772-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...