N-(α)-Ethoxycarbonyl-L-asparagine 97%

L09327-03 L09327-06 L09327-03 L09327-06
AAAL09327-03EA 76.34 USD
CAAAAL09327-03 AAAL09327-03 CAAAAL09327-06 AAAL09327-06
N-(α)-Ethoxycarbonyl-L-asparagine 97%
N-(alpha)-Ethoxycarbonyl-L-asparagine
Formula: C₇H₁₂N₂O₅
MW: 204.18 g/mol
Melting Pt: ca 166 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00082761
CAS Number: 16639-91-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR