4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide 97%

Warning

L09299-03 L09299-03
AAAL09299-03EA 63.9 USD
AAAL09299-03 CAAAAL09299-03
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide 97%
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
Formula: C₈H₈F₃N₃S
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00041291
CAS Number: 20069-30-1
EINECS: 000-000-0
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR