Pentadecafluorooctanoyl chloride 97%
|
Danger
|
L09294-06
L09294-14
L09294-06
L09294-14
AAAL09294-06EA
57.24
USD
CAAAAL09294-06
AAAL09294-06
CAAAAL09294-14
AAAL09294-14
Pentadecafluorooctanoyl chloride 97%
Pentadecafluorooctanoyl chloride
Formula:
C₈ClF₁₅O Boiling Pt: 130…132 °C Density: 1.744 Storage Temperature: Ambient |
MDL Number:
MFCD00013654 CAS Number: 335-64-8 EINECS: 206-394-2 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...