(S)-(-)-α,α-Diphenylprolinol 98%

Warning

Synonyms: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

L09217-06 L09217-06 L09217-03 L09217-03
AAAL09217-03EA 95.22 USD
CAAAAL09217-06 AAAL09217-03 AAAL09217-06 CAAAAL09217-03
(S)-(-)-α,α-Diphenylprolinol 98%
(S)-(-)-α,α-Diphenylprolinol
Formula: C₁₇H₁₉NO
MW: 253.35
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00075506
CAS Number: 112068-01-6

Order Now

Specification Test Results

Optical rotation -57.5° (c=3 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR