4,4'-Bis(2,5-dimethylstyryl)biphenyl ≥99%

L09188-03 L09188-06 L09188-03 L09188-06
AAAL09188-03EA 37.72 USD
AAAL09188-03 CAAAAL09188-03 AAAL09188-06 CAAAAL09188-06
4,4'-Bis(2,5-dimethylstyryl)biphenyl ≥99%
4,4'-Bis(2,5-dimethylstyryl)biphenyl
Formula: C₃₂H₃₀
MW: 414.59 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00051223
CAS Number: 72814-85-8
EINECS: 276-866-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR