DL-Homocysteic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-4-sulfobutanoic acid2-Amino-4-sulphobutanoic acid

L09122-03 L09122-03
AAAL09122-03EA 43.99 USD
CAAAAL09122-03 AAAL09122-03
DL-Homocysteic acid 98%
DL-Homocysteic acid
Formula: C₄H₉NO₅S
MW: 183.18 g/mol
Melting Pt: ca 273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007536
CAS Number: 504-33-6
EINECS: 207-991-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR