Tetrabenzyl pyrophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

L09083-03 L09083-06 L09083-MD L09083-03
AAL09083-06EA 809.99 USD
CAAAAL09083-03 AAL09083-06 AAAL09083-03 CAAAAL09083-MD
Tetrabenzyl pyrophosphate 98%
Tetrabenzyl pyrophosphate
Formula: C₂₈H₂₈O₇P₂
Melting Pt: 58…64 °C
Storage Temperature: Freezer
MDL Number: MFCD00051941
CAS Number: 990-91-0
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR