2-Hydroxy-3-methylbenzhydrazide 98%

Warning

Synonyms: 2-Hydroxy-3-methylbenzohydrazide

L08980-03 L08980-03 L08980-06 L08980-06
AAAL08980-03EA 54.08 USD
CAAAAL08980-03 AAAL08980-03 CAAAAL08980-06 AAAL08980-06
2-Hydroxy-3-methylbenzhydrazide 98%
2-Hydroxy-3-methylbenzhydrazide
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00017052
CAS Number: 30991-42-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR