4-(Methylthio)benzonitrile 98+%

Danger

L08979-22 L08979-14 L08979-14 L08979-22
AAAL08979-14EA 502.46 USD
AAAL08979-14 CAAAAL08979-14 AAAL08979-22 CAAAAL08979-22
4-(Methylthio)benzonitrile 98+%
4-(Methylthio)benzonitrile
Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 272 °C
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00010381
CAS Number: 21382-98-9
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR