1-Chloro-6-phenylhexane 98+%

L08949-03 L08949-MD L08949-MD L08949-03
AAAL08949-03EA 96.68 USD
AAAL08949-03 CAAAAL08949-MD CAAAAL08949-03 AAAL08949-MD
1-Chloro-6-phenylhexane 98+%
1-Chloro-6-phenylhexane
Formula: C₁₂H₁₇Cl
Boiling Pt: 88…91 °C/0.4 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013698
CAS Number: 71434-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR