1-Chloro-7-phenylheptane 98+%

L08817-03 L08817-03 L08817-MD L08817-MD
AAAL08817-03EA 100.86 USD
CAAAAL08817-MD AAAL08817-03 CAAAAL08817-03 AAAL08817-MD
1-Chloro-7-phenylheptane 98+%
1-Chloro-7-phenylheptane
Formula: C₁₃H₁₉Cl
Boiling Pt: 94…96 °C/0.3 mm
Storage Temperature: Ambient
MDL Number: MFCD00039392
CAS Number: 71434-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR