2-Iodo-3-pyridinol 98%

Warning

Synonyms: 3-Hydroxy-2-iodopyridine

L08667-14 L08667-03 L08667-03 L08667-06 L08667-06
AAAL08667-03EA 64.14 USD
CAAAAL08667-14 CAAAAL08667-03 AAAL08667-03 CAAAAL08667-06 AAAL08667-06
2-Iodo-3-pyridinol 98%
2-Iodo-3-pyridinol
Formula: C₅H₄INO
MW: 221.00 g/mol
Melting Pt: 191…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00023421
EINECS: 254-864-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR