4'-Benzyloxy-3'-methoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(4-(Benzyloxy)-3-methoxyphenyl)ethanone
L08430-06
L08430-03
AAAL08430-06EA
200.99
USD
AAAL08430-06
AAL08430-03
4'-Benzyloxy-3'-methoxyacetophenone 98%
4'-Benzyloxy-3'-methoxyacetophenone
Formula:
C₁₆H₁₆O₃ MW: 256.30 g/mol Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017326 CAS Number: 1835-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...