3,5-Dichloro-2-pyridone 98+%

Supplier: Thermo Scientific Chemicals
Warning

L08309-06 L08309-03 L08309-06 L08309-03
AAAL08309-06EA 204.99 USD
AAAL08309-06 CAAAAL08309-06 AAAL08309-03 CAAAAL08309-03
3,5-Dichloro-2-pyridone 98+%
3,5-Dichloro-2-pyridone
Formula: C₅H₃Cl₂NO
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00015401
EINECS: 226-609-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR