2-(2,4,5-Trichlorophenoxy)propionic acid 96%
|
Warning
|
L08136-06
L08136-14
L08136-14
L08136-06
AAAL08136-14EA
375.09
USD
AAAL08136-14
CAAAAL08136-14
CAAAAL08136-06
AAAL08136-06
2-(2,4,5-Trichlorophenoxy)propionic acid 96%
2-(2,4,5-Trichlorophenoxy)propionic acid
Formula:
C₉H₇Cl₃O₃ Melting Pt: 176…180 °C Storage Temperature: Ambient |
EINECS:
202-271-2 UN: 3077 ADR: "9",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...