4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%

Supplier: Thermo Scientific Chemicals

Danger

L08087-03 L08087-03
AAAL08087-03EA 351.27 USD
CAAAAL08087-03 AAAL08087-03
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol 98%
4-[(1R)-2-Amino-1-hydroxyethyl]benzene-1,2-diol
Formula: C₈H₁₁NO₃
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00025592
CAS Number: 51-41-2
EINECS: 200-096-6
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Optical rotation -45° (c=5 in 0.5 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR