3-tert-Butyl-1-methyl-2-pyrazolin-5-one 98%
Synonyms:
5-(tert-Butyl)-2,4-dihydro-2-methyl-3H-pyrazol-3-one
L07849-03
L07849-03
L07849-06
L07849-06
AAAL07849-06EA
240.4
USD
AAAL07849-06
CAAAAL07849-03
AAAL07849-03
CAAAAL07849-06
3-tert-Butyl-1-methyl-2-pyrazolin-5-one 98%
3-tert-Butyl-1-methyl-2-pyrazolin-5-one
Formula:
C₈H₁₄N₂O MW: 154.21 g/mol Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051761 CAS Number: 87031-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...