Trifluorolactic acid 96%

Danger

L07742-03 L07742-06 L07742-03 L07742-06
AAAL07742-06EA 262.57 USD
AAAL07742-06 CAAAAL07742-03 AAAL07742-03 CAAAAL07742-06
Trifluorolactic acid 96%
Trifluorolactic acid
Formula: C₃H₃F₃O₃
MW: 144.05 g/mol
Boiling Pt: 157 °C/123 mm
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00051802
CAS Number: 684-07-1
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR