1-(4-Fluorophenyl)biguanide hydrochloride 98%

Warning

L07330-03 L07330-03 L07330-06 L07330-06
AAAL07330-06EA 99.56 USD
AAAL07330-06 AAAL07330-03 CAAAAL07330-06 CAAAAL07330-03
1-(4-Fluorophenyl)biguanide hydrochloride 98%
1-(4-Fluorophenyl)biguanide hydrochloride
Formula: C₈H₁₁ClFN₅
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00051986
CAS Number: 16018-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR