2,5-Bis(1-naphthyl)-1,3,4-oxadiazole 99%

Warning

L07215-03 L07215-06 L07215-06 L07215-03
AAAL07215-06EA 166.27 USD
AAAL07215-06 AAAL07215-03 CAAAAL07215-03 CAAAAL07215-06
2,5-Bis(1-naphthyl)-1,3,4-oxadiazole 99%
2,5-Bis(1-naphthyl)-1,3,4-oxadiazole
Formula: C₂₂H₁₄N₂O
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00045582
CAS Number: 905-62-4
EINECS: 212-995-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR