Ethyl-5-chlorothiophene-2-carboxylate 98+%

Warning

Synonyms: Ethyl 5-chlorothiophene-2-carboxylate

L06385-06 L06385-06 L06385-03 L06385-14 L06385-03 L06385-14
AAAL06385-03EA 43.98 USD
AAAL06385-03 AAAL06385-14 CAAAAL06385-06 AAAL06385-06 CAAAAL06385-03 CAAAAL06385-14
Ethyl-5-chlorothiophene-2-carboxylate 98+%
Ethyl 5-chlorothiophene-2-carboxylate
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 70…71 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00051732
CAS Number: 5751-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR