1-(4-Methoxyphenyl)cyclohexanecarbonitrile 94%

Warning

L06107-06 L06107-14 L06107-06 L06107-14
AAAL06107-06EA 81.03 USD
CAAAAL06107-14 AAAL06107-06 CAAAAL06107-06 AAAL06107-14
1-(4-Methoxyphenyl)cyclohexanecarbonitrile 94%
1-(4-Methoxyphenyl)cyclohexanecarbonitrile
Formula: C₁₄H₁₇NO
Melting Pt: 43…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00021282
CAS Number: 36263-51-1
EINECS: 252-938-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR