DL-4-Hydroxymandelic acid monohydrate 97%

Danger

Synonyms: 2-Hydroxy-2-(4-hydroxyphenyl)acetic acid hydrateDL-alpha,4-Dihydroxyphenylacetic Acid

L05916-06 L05916-06 L05916-14
AAAL05916-06EA 32.61 USD
AAAL05916-06 CAAAAL05916-06 AAL05916-14
DL-4-Hydroxymandelic acid monohydrate 97%
DL-4-Hydroxymandelic acid monohydrate
Formula: C₈H₁₀O₅
MW: 186,16 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149283
CAS Number: 184901-84-6
EINECS: 214-839-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR