2-Amino-4-chlorobenzothiazole hydrobromide 97%

Warning

L05440-06 L05440-06
AAAL05440-06EA 59.47 USD
CAAAAL05440-06 AAAL05440-06
2-Amino-4-chlorobenzothiazole hydrobromide 97%
2-Amino-4-chlorobenzothiazole hydrobromide
Formula: C₇H₅ClN₂S·HBr
MW: 265.57 g/mol
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00035186
CAS Number: 27058-83-9
EINECS: 248-194-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR