Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibenzyl phthalate 97%

Supplier: Alfa Aesar

L04985-06 L04985-06 L04985-14 L04985-14
AAAL04985-06EA 71.28 USD InStock
CAAAAL04985-06 AAAL04985-06 CAAAAL04985-14 AAAL04985-14
Dibenzyl phthalate 97%
Dibenzyl phthalate


Formula: C₂₂H₁₈O₄
MW: 346,39
Boiling Pt: 276-278°C/15mm
Melting Pt: 40-42°C
Storage Temperature: Ambient
MDL Number: MFCD00014437
CAS Number: 523-31-9
EINECS: 208-344-5
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty