Allyl phenyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Warning

L04771-18 L04771-18 L04771-09 L04771-09
AAAL04771-18EA 296.71 USD
CAAAAL04771-18 AAAL04771-18 CAAAAL04771-09 AAAL04771-09
Allyl phenyl sulfide 97%
Allyl phenyl sulphide
Formula: C₉H₁₀S
Boiling Pt: 223…225 °C
Density: 1,024
Flash Pt: >98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014957
CAS Number: 5296-64-0
EINECS: 226-142-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR