1,3-Bis(chloromethyl)tetramethyldisiloxane 97%

Warning

L04436-09 L04436-18 L04436-18 L04436-09
AAAL04436-09EA 47.45 USD
CAAAAL04436-18 AAAL04436-09 AAAL04436-18 CAAAAL04436-09
1,3-Bis(chloromethyl)tetramethyldisiloxane 97%
1,3-Bis(chloromethyl)tetramethyldisiloxane
Formula: C₆H₁₆Cl₂OSi₂
MW: 231.26 g/mol
Boiling Pt: 92…93 °C/20 mm
Density: 1.050
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000877
CAS Number: 2362-10-9
EINECS: 219-109-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR