2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate 99%

L04024-14 L04024-14 L04024-22 L04024-22
AAAL04024-22EA 56.33 USD
CAAAAL04024-22 CAAAAL04024-14 AAAL04024-22 AAAL04024-14
2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate 99%
2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate
Formula: C₁₃H₁₉NO₂·HCl·xH₂O
MW: 257.76 g/mol (anhy)
Melting Pt: 181…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00150097
CAS Number: 33625-43-3
EINECS: 251-600-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR