Monoethyl glutarate 97%

Warning

Synonyms: Glutaric Acid Monoethyl Ester5-Ethoxy-5-oxopentanoic acid

L03941-06 L03941-14 L03941-14 L03941-06
AAAL03941-14EA 343.63 USD
CAAAAL03941-14 CAAAAL03941-06 AAAL03941-14 AAAL03941-06
Monoethyl glutarate 97%
Monoethyl glutarate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 118…120 °C/1.5 mm
Storage Temperature: Ambient
MDL Number: MFCD00014379
CAS Number: 1070-62-8
EINECS: 213-977-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR