1-(1-Acetyl-1H-indol-3-yl)-1-ethanone 97%

Synonyms: 1,3-Diacetyl-1H-indole1,3-Diacetylindole

L03698-06 L03698-03 L03698-06 L03698-03
AAAL03698-06EA 127.65 USD
AAAL03698-06 AAAL03698-03 CAAAAL03698-06 CAAAAL03698-03
1-(1-Acetyl-1H-indol-3-yl)-1-ethanone 97%
1-(1-Acetyl-1H-indol-3-yl)-1-ethanone
Formula: C₁₂H₁₁NO₂
MW: 201.23 g/mol
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00039692
CAS Number: 17537-64-3
EINECS: 241-532-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR