4-Bromo-N,N-diethylaniline 97%

Supplier: Thermo Scientific Chemicals

Danger

L03619-09 L03619-09
AAAL03619-09EA 86.4 USD
CAAAAL03619-09 AAAL03619-09
4-Bromo-N,N-diethylaniline 97%
4-Bromo-N,N-diethylaniline
Formula: C₁₀H₁₄BrN
MW: 228.13 g/mol
Boiling Pt: 269…271 °C
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00013530
EINECS: 218-140-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR