5-(Benzyloxy)-2-nitrotoluene 98%

Synonyms: 4-(Benzyloxy)-2-methyl-1-nitrobenzene

L03538-06 L03538-06 L03538-03 L03538-03
AAAL03538-06EA 104.67 USD
CAAAAL03538-03 AAAL03538-06 CAAAAL03538-06 AAAL03538-03
5-(Benzyloxy)-2-nitrotoluene 98%
5-(Benzyloxy)-2-nitrotoluene
Formula: C₁₄H₁₃NO₃
MW: 243.26 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00024269
CAS Number: 22424-58-4
EINECS: 244-988-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR