Thiodisuccinic acid 98+%

Warning

Synonyms: 2,2'-Thiodisuccinic acid

L03222-14 L03222-14 L03222-06 L03222-06
AAAL03222-06EA 34.97 USD
CAAAAL03222-06 AAAL03222-06 CAAAAL03222-14 AAAL03222-14
Thiodisuccinic acid 98+%
Thiodisuccinic acid
Formula: C₈H₁₀O₈S
MW: 266.22 g/mol
Melting Pt: ca 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00020502
CAS Number: 4917-76-4
EINECS: 225-544-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR