Ethyl-3,5-dimethylbenzoate 98+%

Warning

Synonyms: Ethyl 3,5-dimethylbenzoate

L02890-14 L02890-06 L02890-06 L02890-14
AAAL02890-14EA 266.4 USD
CAAAAL02890-14 AAAL02890-14 CAAAAL02890-06 AAAL02890-06
Ethyl-3,5-dimethylbenzoate 98+%
Ethyl 3,5-dimethylbenzoate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 120 °C/10 mm
Density: 1,113
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015440
CAS Number: 21239-29-2
EINECS: 244-286-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR