Ethyl-2,4-dimethylbenzoate 98+%

Synonyms: Ethyl 2,4-dimethylbenzoate

L02759-06 L02759-14 L02759-06 L02759-14
AAAL02759-14EA 185.37 USD
CAAAAL02759-06 AAAL02759-14 CAAAAL02759-14 AAAL02759-06
Ethyl-2,4-dimethylbenzoate 98+%
Ethyl 2,4-dimethylbenzoate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015438
CAS Number: 33499-42-2
EINECS: 251-544-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR