1-(2-Thienyl)-1-propanone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Propionylthiophene

L02318-14 L02318-14 L02318-06 L02318-06 L02318-22 L02318-22
AAAL02318-06EA 26.99 USD
AAAL02318-06 CAAAAL02318-06 AAAL02318-14 CAAAAL02318-14 AAAL02318-22 CAAAAL02318-22
1-(2-Thienyl)-1-propanone 98%
1-(2-Thienyl)-1-propanone
Formula: C₇H₈OS
MW: 140.20 g/mol
Boiling Pt: 228 °C
Density: 1.124
Flash Pt: 100 °C(212 °F)
Storage Temperature: Ambient
EINECS: 237-182-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR