1-(3-Methoxyphenyl)piperazine dihydrochloride 97%

Warning

L02164-06 L02164-06 L02164-14 L02164-14
AAAL02164-06EA 79.23 USD
AAAL02164-06 CAAAAL02164-06 CAAAAL02164-14 AAAL02164-14
1-(3-Methoxyphenyl)piperazine dihydrochloride 97%
1-(3-Methoxyphenyl)piperazine dihydrochloride
Formula: C₁₁H₁₈Cl₂N₂O
Melting Pt: ca 214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035285
CAS Number: 6968-76-9
EINECS: 230-189-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR