(4-Pentenyl)triphenylphosphonium bromide 97%

Warning

L01950-03 L01950-06 L01950-14 L01950-03 L01950-06 L01950-14
AAAL01950-06EA 211.31 USD
CAAAAL01950-03 CAAAAL01950-14 AAAL01950-06 AAAL01950-03 AAAL01950-14 CAAAAL01950-06
(4-Pentenyl)triphenylphosphonium bromide 97%
(4-Pentenyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrP
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00050215
CAS Number: 56771-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR