Ethyl-3-(morpholino)propionate 97%

Warning

Synonyms: 4-[2-(Ethoxycarbonyl)ethyl]morpholine3-(Morpholino)propionic Acid Ethyl Ester

L01734-14 L01734-06 L01734-06 L01734-14
AAAL01734-06EA 74.32 USD
AAAL01734-06 AAAL01734-14 CAAAAL01734-06 CAAAAL01734-14
Ethyl-3-(morpholino)propionate 97%
Ethyl-3-(morpholino)propionate
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014622
CAS Number: 20120-24-5
EINECS: 243-526-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR