2-(4-Ethylphenoxy)acetic acid 97%

Warning

L01102-06 L01102-06
AAAL01102-06EA 49.45 USD
AAAL01102-06 CAAAAL01102-06
2-(4-Ethylphenoxy)acetic acid 97%
2-(4-Ethylphenoxy)acetic acid
Formula: C₁₀H₁₂O₃
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00014366
CAS Number: 24431-27-4
EINECS: 246-245-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR