1-(4-Formylphenyl)pyrrolidine ≥98%

Warning

Synonyms: 4-(1-Pyrrolidino)benzaldehyde4-(1-Pyrrolidinyl)benzaldehyde

L00966-06 L00966-14 L00966-06
AAAL00966-06EA 77.13 USD
AAAL00966-06 CAAAAL00966-14 CAAAAL00966-06
1-(4-Formylphenyl)pyrrolidine ≥98%
1-(4-Formylphenyl)pyrrolidine
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015469
CAS Number: 51980-54-2
EINECS: 257-570-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR